Gene Transcripts

Gene Symbol
SCARF2
Entrez Gene ID
91179
Gene Name
scavenger receptor class F member 2
Gene Aliases
NSR1, SREC-II, SREC2, SRECRP-1, VDEGS
Chromosome Location
Chr. 22: 20424584 - 20437859 on Build GRCh38
UniGene ID
Hs.474251
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153334.6 NP_699165.3 10 1794
NM_182895.4 NP_878315.2 10 1779
XM_017029065.1 XP_016884554.1 10 1825
GenBank mRNA AB024433.1 - 10 1745
AB052951.1 - 10 1685
AF522196.1 - 10 1794
BC009326.2 - 7 862

View Full Product Details