Gene Transcripts

Gene Symbol
LPCAT4
Entrez Gene ID
254531
Gene Name
lysophosphatidylcholine acyltransferase 4
Gene Aliases
AGPAT7, AYTL3, LPAAT-eta, LPEAT2
Chromosome Location
Chr. 15: 34358888 - 34367196 on Build GRCh38
UniGene ID
Hs.352614
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153613.2 NP_705841.2 2 336
XM_006720454.2 XP_006720517.1 2 338
XM_011521422.1 XP_011519724.1 2 318
GenBank mRNA AF529233.1 - 2 187
AF542964.1 - 2 420
AK001119.1 - 1 382
AK290273.1 - 2 307
AK308910.1 - 2 318
AL832237.1 - 2 333
AY734233.1 - 2 356
BC048430.1 - 2 338
BC092463.1 - 2 333 30520144
BC144236.1 - 2 264

View Full Product Details