Gene Transcripts
- Gene Symbol
- LVRN
- Entrez Gene ID
- 206338
- Gene Name
- laeverin
- Gene Aliases
- APQ, AQPEP, TAQPEP
- Chromosome Location
- Chr. 5: 115962440 - 116027602 on Build GRCh38
- UniGene ID
- Hs.556880
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173800.4 |
NP_776161.3
|
9, 10 |
1806 |
|
XM_011543263.2 |
XP_011541565.1
|
9, 10 |
1820 |
|
XM_011543264.2 |
XP_011541566.1
|
9, 10 |
1820 |
|
GenBank mRNA |
AK075131.1 |
- |
4, 5 |
341 |
|
AK291655.1 |
- |
9, 10 |
1820 |
|
AY560010.1 |
- |
9, 10 |
1694 |
|
BC036440.1 |
- |
4, 5 |
459 |
|
BC045809.1 |
- |
4, 5 |
434 |
|
BC060869.1 |
- |
10, 11 |
1733 |
30346049 |
BC068560.1 |
- |
9, 10 |
1806 |
30341970 |
BC070028.1 |
- |
4, 5 |
454 |
|
BC094716.1 |
- |
4, 5 |
454 |
|
BC109022.1 |
- |
9, 10 |
1670 |
40008501 |
BC109023.1 |
- |
9, 10 |
1670 |
40008502 |
View Full Product Details