Gene Transcripts

Gene Symbol
TAS2R38
Entrez Gene ID
5726
Gene Name
taste 2 receptor member 38
Gene Aliases
PTC, T2R38, T2R61
Chromosome Location
Chr. 7: 141972631 - 141973773 on Build GRCh38
UniGene ID
Hs.647085
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176817.4 NP_789787.4 1 776
GenBank mRNA BC104933.1 - 1 776
BC104937.1 - 1 776 8143940

View Full Product Details