Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012228.3 | NP_036360.3 | 2 | 274 | |
XM_011519426.2 | XP_011517728.1 | 2 | 208 | ||
GenBank mRNA | AF122004.1 | - | 2 | 191 | |
AF151889.1 | - | 2 | 222 | ||
BC018030.1 | - | 2 | 269 | 4799583 | |
BC066305.1 | - | 2 | 188 | ||
BC117471.1 | - | 2 | 323 | 40126022 | |
BC130380.1 | - | 2 | 323 | 40146411 |