Gene Transcripts
- Gene Symbol
- MS4A4A
- Entrez Gene ID
- 51338
- Gene Name
- membrane spanning 4-domains A4A
- Gene Aliases
- 4SPAN1, CD20-L1, CD20L1, HDCME31P, MS4A4, MS4A7
- Chromosome Location
- Chr. 11: 60280541 - 60308972 on Build GRCh38
- UniGene ID
- Hs.325960
- Species
- Homo sapiens
- Gene Symbol
- MS4A4E
- Entrez Gene ID
- 643680
- Gene Name
- membrane spanning 4-domains A4E
- Gene Aliases
- Chromosome Location
- Chr. 11: 60201253 - 60243124 on Build GRCh38
- UniGene ID
- Hs.559934
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB231725.1 |
- |
1 |
19 |
|
View Full Product Details