Gene Transcripts

Gene Symbol
ZGRF1
Entrez Gene ID
55345
Gene Name
zinc finger GRF-type containing 1
Gene Aliases
C4orf21
Chromosome Location
Chr. 4: 112539333 - 112636995 on Build GRCh38
UniGene ID
Hs.380346
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018392.4 NP_060862.3 25, 26 5983
XM_005263115.3 XP_005263172.1 25, 26 6038
XM_011532091.2 XP_011530393.1 24, 25 5932
XM_011532092.2 XP_011530394.1 24, 25 5863
XM_011532093.2 XP_011530395.1 24, 25 5809
XM_011532094.2 XP_011530396.1 23, 24 5725
XM_011532097.2 XP_011530399.1 18, 19 3085
XM_011532098.2 XP_011530400.1 16, 17 2759
XM_011532099.2 XP_011530401.1 15, 16 2718
XM_017008369.1 XP_016863858.1 24, 25 5978
XM_017008370.1 XP_016863859.1 23, 24 5758
XM_017008371.1 XP_016863860.1 22, 23 5638
XM_017008372.1 XP_016863861.1 22, 23 5017
XM_017008373.1 XP_016863862.1 21, 22 4954
GenBank mRNA AK090556.1 - 7, 8 1618
AK126054.1 - 14, 15 2367
AL137700.1 - 10, 11 1430
BX647594.1 - 13, 14 2338

View Full Product Details