Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318942.1 | NP_001305871.1 | 2 | 165 | |
NM_001318943.1 | NP_001305872.1 | 2 | 546 | ||
NM_014623.3 | NP_055438.1 | 2 | 208 | ||
XM_005249121.3 | XP_005249178.1 | 2 | 273 | ||
XM_017010868.1 | XP_016866357.1 | 2 | 104 | ||
GenBank mRNA | AK309759.1 | - | 2 | 106 | |
BC001754.1 | - | 2 | 166 | 3355279 | |
BM450289.1 | - | 109 | |||
BM552852.1 | - | 2 | 524 | ||
BQ232807.1 | - | 2 | 208 | ||
BT009831.1 | - | 2 | 107 | ||
HY039733.1 | - | 2 | 165 | ||
L10400.1 | - | 2 | 141 | ||
M27937.1 | - | 2 | 141 |