Gene Transcripts

Gene Symbol
Lgals5
Entrez Gene ID
25475
Gene Name
lectin, galactose binding, soluble 5
Gene Aliases
Chromosome Location
Chr. 10: 66226015 - 66229293 on Build Rnor_6.0
UniGene ID
Rn.64588
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012976.2 NP_037108.1 4 384
GenBank mRNA L21711.1 - 4 393
L36862.1 - 4 384

View Full Product Details