Gene Transcripts
- Gene Symbol
- Serpina3c
- Entrez Gene ID
- 24794
- Gene Name
- serine (or cysteine) proteinase inhibitor, clade A, member 3C
- Gene Aliases
- CPi-21, GHR-P63, Kbp, Klkbp, SPI-2, SPI-2.3, SPI2, Serpina3k, Spin2, Spin2b
- Chromosome Location
- Chr. 6: 127996162 - 128003435 on Build Rnor_6.0
- UniGene ID
- Rn.91257
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012657.2 |
NP_036789.2
|
2, 3 |
733 |
|
XM_006240458.2 |
XP_006240520.1
|
2, 3 |
776 |
|
GenBank mRNA |
BC062236.1 |
- |
2, 3 |
737 |
|
CO560755.1 |
- |
1, 2 |
228 |
|
D00751.1 |
- |
2, 3 |
686 |
|
FQ209380.1 |
- |
2, 3 |
734 |
|
FQ209577.1 |
- |
2, 3 |
735 |
|
FQ210286.1 |
- |
2, 3 |
738 |
|
FQ218085.1 |
- |
2, 3 |
735 |
|
FQ218647.1 |
- |
1, 2 |
410 |
|
FQ219296.1 |
- |
2, 3 |
738 |
|
FQ219312.1 |
- |
2, 3 |
735 |
|
M14320.1 |
- |
1, 2 |
228 |
|
M15916.1 |
- |
2, 3 |
698 |
|
X05348.1 |
- |
2, 3 |
720 |
|
X16358.1 |
- |
2, 3 |
734 |
|
View Full Product Details