Gene Transcripts

Gene Symbol
Smpd5
Entrez Gene ID
100503915
Gene Name
sphingomyelin phosphodiesterase 5
Gene Aliases
ENSMUSG00000071724, Gm10345, MA-nSMase
Chromosome Location
Chr. 15: 76293869 - 76296896 on Build GRCm38
UniGene ID
Mm.393455
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195537.1 NP_001182466.1 1 666
GenBank mRNA GU144514.1 - 1 666

View Full Product Details