Gene Transcripts

Gene Symbol
Mki67
Entrez Gene ID
17345
Gene Name
antigen identified by monoclonal antibody Ki 67
Gene Aliases
D630048A14Rik, Ki-67, Ki67
Chromosome Location
Chr. 7: 135689782 - 135716408 on Build GRCm38
UniGene ID
Mm.4078
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081117.2 NP_001074586.2 12 8123
GenBank mRNA AK133964.1 - 1 646
BC053453.1 - 1 749
X82786.1 - 13 7508

View Full Product Details