Gene Transcripts

Gene Symbol
Larp4
Entrez Gene ID
207214
Gene Name
La ribonucleoprotein domain family, member 4
Gene Aliases
DXErtd793, DXErtd793e
Chromosome Location
Chr. 15: 99970065 - 100016358 on Build GRCm38
UniGene ID
Mm.28811
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024526.2 NP_001019697.2 16 3958
NM_001080948.2 NP_001074417.1 16 3971
NM_001284521.1 NP_001271450.1 16 3945
NM_001284522.1 NP_001271451.1 16 3968
NM_001284523.1 NP_001271452.1 16 3961
GenBank mRNA AK052358.1 - 1 219
AK079933.1 - 1 207
AK139636.1 - 16 3967
AK139909.1 - 16 3811
AK141743.1 - 1 1751

View Full Product Details