Gene Transcripts

Gene Symbol
Lilra5
Entrez Gene ID
232801
Gene Name
leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 5
Gene Aliases
E030017K20, EG232801, Gm4878, Lilrc1
Chromosome Location
Chr. 7: 4237643 - 4243690 on Build GRCm38
UniGene ID
Mm.258061
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081239.2 NP_001074708.2 6 2003
GenBank mRNA AK086983.1 - 3 1417

View Full Product Details