Gene Transcripts

Gene Symbol
Nbeal2
Entrez Gene ID
235627
Gene Name
neurobeachin-like 2
Gene Aliases
1110014F23Rik, BC042396, mKIAA0540
Chromosome Location
Chr. 9: 110624789 - 110654161 on Build GRCm38
UniGene ID
Mm.335481
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183276.2 NP_899099.2 50 7851
GenBank mRNA AK129158.1 - 25 4119
BC157956.1 - 50 7832 9007284
BC172118.1 - 49 7751 9054404

View Full Product Details