Gene Transcripts

Gene Symbol
Trim63
Entrez Gene ID
433766
Gene Name
tripartite motif-containing 63
Gene Aliases
MuRF1, RF1, Rnf28
Chromosome Location
Chr. 4: 134315088 - 134329629 on Build GRCm38
UniGene ID
Mm.331961
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039048.2 NP_001034137.2 9 1501
GenBank mRNA AJ278734.1 - 2 375
AK209687.1 - 1 109

View Full Product Details