Gene Transcripts

Gene Symbol
Vmn2r82
Entrez Gene ID
624845
Gene Name
vomeronasal 2, receptor 82
Gene Aliases
EG624845
Chromosome Location
Chr. 10: 79356591 - 79396766 on Build GRCm38
UniGene ID
Mm.469866
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101572.1 NP_001095042.1 6 1753

View Full Product Details