Gene Transcripts

Gene Symbol
Inafm1
Entrez Gene ID
66300
Gene Name
InaF motif containing 1
Gene Aliases
2610014I16Rik, Prr24
Chromosome Location
Chr. 7: 16272013 - 16273692 on Build GRCm38
UniGene ID
Mm.218698
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136270.1 NP_001129742.1 1 169
GenBank mRNA CX734215.1 - 1 199

View Full Product Details