Gene Transcripts

Gene Symbol
Lelp1
Entrez Gene ID
69332
Gene Name
late cornified envelope-like proline-rich 1
Gene Aliases
1700012F11Rik
Chromosome Location
Chr. 3: 92134994 - 92142754 on Build GRCm38
UniGene ID
Mm.120271
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027042.1 NP_081318.1 2 180
GenBank mRNA AK005903.1 - 2 171

View Full Product Details