Gene Transcripts

Gene Symbol
Lce3e
Entrez Gene ID
69514
Gene Name
late cornified envelope 3E
Gene Aliases
2310001H18Rik
Chromosome Location
Chr. 3: 92967061 - 92968281 on Build GRCm38
UniGene ID
Mm.440092
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001254725.1 NP_001241654.1 2 578
GenBank mRNA AK009065.1 - 1 276

View Full Product Details