Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027526.1 | NP_081802.1 | 10 | 1233 | |
GenBank mRNA | AK158756.1 | - | 11 | 1259 | |
BC138284.1 | - | 10 | 1199 | 8861303 | |
BC145232.1 | - | 10 | 1226 | 9053784 | |
BC145752.1 | - | 10 | 1199 | 40131055 |