Gene Transcripts

Gene Symbol
Rasgef1a
Entrez Gene ID
70727
Gene Name
RasGEF domain family, member 1A
Gene Aliases
6330404M18Rik, AI835194
Chromosome Location
Chr. 6: 118011466 - 118091546 on Build GRCm38
UniGene ID
Mm.87660
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027526.1 NP_081802.1 7 871
GenBank mRNA AK018120.1 - 6 702
AK158756.1 - 8 897
BC138284.1 - 7 837 8861303
BC145232.1 - 7 864 9053784
BC145752.1 - 7 837 40131055

View Full Product Details