Gene Transcripts

Gene Symbol
Lst1
Entrez Gene ID
64569
Gene Name
leukocyte specific transcript 1
Gene Aliases
B144
Chromosome Location
Chr. 20: 5175291 - 5179353 on Build Rnor_6.0
UniGene ID
Rn.91085
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022634.2 NP_072156.2 5 291
XM_006256078.2 XP_006256140.1 5 432
XM_006256079.2 XP_006256141.1 5 432
XM_006256080.2 XP_006256142.1 4 379
GenBank mRNA AF208230.1 - 5 291
FQ222284.1 - 4 289

View Full Product Details