Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001111098.1 | NP_001104568.1 | 3 | 332 | |
NM_030857.2 | NP_110484.1 | 3 | 395 | ||
XM_006237834.2 | XP_006237896.1 | 3 | 711 | ||
GenBank mRNA | AF000300.1 | - | 3 | 240 | |
AF000301.1 | - | 3 | 332 | ||
AF000302.1 | - | 3 | 330 | ||
L14951.1 | - | 3 | 398 |