Gene Transcripts
- Gene Symbol
- Picalm
- Entrez Gene ID
- 89816
- Gene Name
- phosphatidylinositol binding clathrin assembly protein
- Gene Aliases
- Calm
- Chromosome Location
- Chr. 1: 154377234 - 154458922 on Build Rnor_6.0
- UniGene ID
- Rn.24871
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053554.2 |
NP_446006.1
|
4, 5 |
483 |
|
XM_006229669.2 |
XP_006229731.1
|
4, 5 |
753 |
|
XM_006229670.2 |
XP_006229732.1
|
4, 5 |
753 |
|
XM_006229671.2 |
XP_006229733.1
|
4, 5 |
754 |
|
XM_006229672.2 |
XP_006229734.1
|
4, 5 |
753 |
|
XM_006229673.2 |
XP_006229735.1
|
4, 5 |
753 |
|
XM_006229674.2 |
XP_006229736.1
|
4, 5 |
753 |
|
XM_006229675.2 |
XP_006229737.1
|
4, 5 |
753 |
|
XM_006229676.2 |
XP_006229738.1
|
4, 5 |
752 |
|
XM_006229677.2 |
XP_006229739.1
|
4, 5 |
752 |
|
XM_006229678.2 |
XP_006229740.1
|
4, 5 |
752 |
|
XM_006229679.2 |
XP_006229741.1
|
4, 5 |
752 |
|
XM_006229680.2 |
XP_006229742.1
|
4, 5 |
752 |
|
XM_006229681.2 |
XP_006229743.1
|
4, 5 |
752 |
|
XM_006229682.2 |
XP_006229744.1
|
4, 5 |
752 |
|
XM_006229683.2 |
XP_006229745.1
|
4, 5 |
752 |
|
XM_006229684.2 |
XP_006229746.1
|
4, 5 |
752 |
|
XM_006229685.2 |
XP_006229747.1
|
4, 5 |
752 |
|
GenBank mRNA |
AF041373.1 |
- |
4, 5 |
484 |
|
AF041374.1 |
- |
4, 5 |
484 |
|
AY699717.1 |
- |
4, 5 |
459 |
|
AY706215.1 |
- |
4, 5 |
459 |
|
AY724477.1 |
- |
4, 5 |
477 |
|
BC100142.1 |
- |
4, 5 |
523 |
7456159 |
View Full Product Details