Gene Transcripts

Gene Symbol
Ppp2r2c
Entrez Gene ID
117256
Gene Name
protein phosphatase 2, regulatory subunit B, gamma
Gene Aliases
Pr52
Chromosome Location
Chr. 14: 78679214 - 78763175 on Build Rnor_6.0
UniGene ID
Rn.54550
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057116.1 NP_476457.1 1325
GenBank mRNA D38261.1 - 1325

View Full Product Details