Gene Transcripts

Gene Symbol
Mylk2
Entrez Gene ID
117558
Gene Name
myosin light chain kinase 2
Gene Aliases
Klmc, Mlck
Chromosome Location
Chr. 3: 148386185 - 148397851 on Build Rnor_6.0
UniGene ID
Rn.81191
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057209.1 NP_476557.1 2 503
GenBank mRNA J03886.1 - 2 503

View Full Product Details