Gene Transcripts

Gene Symbol
Rnf39
Entrez Gene ID
171387
Gene Name
ring finger protein 39
Gene Aliases
Hzfw1, Lirf
Chromosome Location
Chr. 20: 2098375 - 2103864 on Build Rnor_6.0
UniGene ID
Rn.144662
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134374.2 NP_599201.1 3 604
GenBank mRNA AB070897.1 - 3 604

View Full Product Details