Gene Transcripts

Gene Symbol
Phrf1
Entrez Gene ID
245925
Gene Name
PHD and ring finger domains 1
Gene Aliases
Chromosome Location
Chr. 1: 214215677 - 214248906 on Build Rnor_6.0
UniGene ID
Rn.10530
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139093.1 NP_620793.1 8, 9 1017
XM_006230510.2 XP_006230572.1 9, 10 1185
XM_006230511.2 XP_006230573.1 9, 10 1143
XM_006230513.2 XP_006230575.1 9, 10 1182
XM_006230514.2 XP_006230576.1 9, 10 1185
XM_006230515.2 XP_006230577.1 9, 10 1185
XM_006230516.2 XP_006230578.1 9, 10 1185
XM_006230517.2 XP_006230579.1 9, 10 1182
GenBank mRNA U49057.1 - 8, 9 1017

View Full Product Details