Gene Transcripts

Gene Symbol
Pcp4
Entrez Gene ID
25510
Gene Name
Purkinje cell protein 4
Gene Aliases
PEPZ19, pep-19
Chromosome Location
Chr. 11: 36851038 - 36912231 on Build Rnor_6.0
UniGene ID
Rn.9736
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AJ493658.1 - 4 253
BC059147.1 - 3 133 6888437
BG665749.1 - 3 132
BG666168.1 - 3 123
CB613293.1 - 3 128
M24852.1 - 3 120

View Full Product Details