Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008519.2 | NP_032545.1 | 2 | 536 | |
GenBank mRNA | AF044030.1 | - | 2 | 532 | |
AF110104.1 | - | 2 | 483 | ||
AK010895.1 | - | 2 | 522 | ||
AK155181.1 | - | 2 | 537 | ||
BC064063.1 | - | 2 | 467 | 874601 |