Gene Transcripts

Gene Symbol
Npc1
Entrez Gene ID
18145
Gene Name
Niemann-Pick type C1
Gene Aliases
A430089E03Rik, C85354, D18Ertd139e, D18Ertd723e, lcsd, nmf164, spm
Chromosome Location
Chr. 18: 12189694 - 12236386 on Build GRCm38
UniGene ID
Mm.3484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008720.2 NP_032746.2 2 273
GenBank mRNA AF003348.1 - 2 250
AK040367.1 - 2 246
AK044878.1 - 2 214
AK087174.1 - 2 228
AK154282.1 - 2 193
AK155379.1 - 2 193
AK170880.1 - 2 214
AK171371.1 - 2 214
BC052437.1 - 2 273 6405214
BC054539.1 - 2 273 6405214

View Full Product Details