Gene Transcripts
- Gene Symbol
- Pbx1
- Entrez Gene ID
- 18514
- Gene Name
- pre B cell leukemia homeobox 1
- Gene Aliases
- 2310056B04Rik, D230003C07Rik, Pbx-1
- Chromosome Location
- Chr. 1: 168119364 - 168432264 on Build GRCm38
- UniGene ID
- Mm.43358
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001291508.1 |
NP_001278437.1
|
1, 2 |
859 |
|
NM_001291509.1 |
NP_001278438.1
|
1, 2 |
859 |
|
NM_008783.3 |
NP_032809.1
|
1, 2 |
859 |
|
NM_183355.3 |
NP_899198.1
|
1, 2 |
859 |
|
GenBank mRNA |
AB221563.1 |
- |
1, 2 |
194 |
|
AF020196.1 |
- |
1, 2 |
550 |
|
AK037006.1 |
- |
1, 2 |
823 |
|
AK045252.1 |
- |
1, 2 |
858 |
|
AK084143.1 |
- |
1, 2 |
351 |
|
AK144346.1 |
- |
1, 2 |
861 |
|
AK144446.1 |
- |
1, 2 |
861 |
|
BC002244.1 |
- |
1, 2 |
85 |
3492658 |
BC054376.1 |
- |
1, 2 |
820 |
|
BC058390.1 |
- |
1, 2 |
587 |
5701148 |
HM369110.1 |
- |
1, 2 |
194 |
|
L27453.1 |
- |
1, 2 |
238 |
|
View Full Product Details