Gene Transcripts

Gene Symbol
Map2k3
Entrez Gene ID
26397
Gene Name
mitogen-activated protein kinase kinase 3
Gene Aliases
AW212142, MEK3, MKK3, Prkmk3, mMKK3b
Chromosome Location
Chr. 11: 60932057 - 60952803 on Build GRCm38
UniGene ID
Mm.18494
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008928.4 NP_032954.1 12 2129
GenBank mRNA AK008141.1 - 12 2132
AK011002.1 - 12 2130
AK150067.1 - 12 2130
AK159379.1 - 12 2129
AK166255.1 - 12 2131
AK166507.1 - 12 2130
AK167442.1 - 12 2133
BC007467.1 - 11 2068 3497714

View Full Product Details