Gene Transcripts
- Gene Symbol
- Rbm4
- Entrez Gene ID
- 19653
- Gene Name
- RNA binding motif protein 4
- Gene Aliases
- 4921506I22Rik, Lark1, Mlark, Rbm4a, lark
- Chromosome Location
- Chr. 19: 4784293 - 4794009 on Build GRCm38
- UniGene ID
- Mm.426069
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U89506.1 |
- |
5 |
1291 |
|
View Full Product Details