Gene Transcripts

Gene Symbol
OSBPL1A
Entrez Gene ID
114876
Gene Name
oxysterol binding protein like 1A
Gene Aliases
ORP-1, ORP1, OSBPL1B
Chromosome Location
Chr. 18: 24162045 - 24397882 on Build GRCh38
UniGene ID
Hs.370725
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242508.1 NP_001229437.1 4 388
NM_018030.4 NP_060500.3 2 757
NM_080597.3 NP_542164.2 16 1644
XM_006722380.3 XP_006722443.1 15 1557
XM_006722382.3 XP_006722445.1 16 1657
XM_017025530.1 XP_016881019.1 16 1936
XM_017025531.1 XP_016881020.1 15 1429
XM_017025533.1 XP_016881022.1 2 240
XM_017025534.1 XP_016881023.1 2 2768
GenBank mRNA AF274714.1 - 2 757
AF323726.2 - 16 1601
AF392449.1 - 2 378
AF392450.1 - 16 1601
AK001079.1 - 3 285
AK096358.1 - 16 1644
AK292895.1 - 17 1576
AK301862.1 - 4 388
AK314552.1 - 2 208
BC063420.1 - 2 443
BC098384.1 - 2 436
BC136452.1 - 16 1541 9020441
BC144117.1 - 16 1541 9052637
BX647893.1 - 10 3320

View Full Product Details