Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010879.3 | NP_035009.3 | 5 | 2576 | |
GenBank mRNA | AK014949.1 | - | 5 | 2572 | |
AK051665.1 | - | 5 | 2577 | ||
BC011071.1 | - | 3 | 2139 | 3673483 | |
BC034255.1 | - | 5 | 2315 | 3157491 |