Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159561.1 | NP_001153033.1 | 2, 3 | 458 | |
NM_011028.2 | NP_035158.2 | 2, 3 | 458 | ||
GenBank mRNA | AB010883.1 | - | 2, 3 | 343 | |
AK054195.1 | - | 411 | |||
AK081308.1 | - | 2, 3 | 334 | ||
AK138541.1 | - | 2, 3 | 310 | ||
BC117713.2 | - | 2, 3 | 320 | 40088045 |