Gene Transcripts

Gene Symbol
Robo3
Entrez Gene ID
19649
Gene Name
roundabout homolog 3 (Drosophila)
Gene Aliases
Rbig1, Rig-1, Rig1, Robo3a, Robo3b
Chromosome Location
Chr. 9: 37416045 - 37433408 on Build GRCm38
UniGene ID
Mm.212826
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164767.1 NP_001158239.1 3 806
GenBank mRNA AF060570.1 - 2 792

View Full Product Details