Gene Transcripts

Gene Symbol
Tuft1
Entrez Gene ID
22156
Gene Name
tuftelin 1
Gene Aliases
Chromosome Location
Chr. 3: 94612753 - 94658872 on Build GRCm38
UniGene ID
Mm.10214
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011656.3 NP_035786.1 2 202
GenBank mRNA AF002860.1 - 2 96
AF047704.1 - 2 136
AK132488.1 - 2 134
BC019213.1 - 2 133 5064658

View Full Product Details