Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011942.1 | NP_036072.1 | 10 | 776 | |
GenBank mRNA | AB009653.1 | - | 10 | 776 | |
AK003689.1 | - | 10 | 743 | ||
AK075590.1 | - | 10 | 768 | ||
AK089112.1 | - | 10 | 768 | ||
AK167478.1 | - | 10 | 763 | ||
BC068120.1 | - | 10 | 802 | 5714650 |