Gene Transcripts

Gene Symbol
Reg3d
Entrez Gene ID
30053
Gene Name
regenerating islet-derived 3 delta
Gene Aliases
INGAP, Ingaprp
Chromosome Location
Chr. 6: 78375874 - 78378865 on Build GRCm38
UniGene ID
Mm.33691
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161741.1 NP_001155213.1 4, 5 380
NM_013893.2 NP_038921.2 5, 6 518
GenBank mRNA AB028625.1 - 5, 6 524
AB035204.1 - 5, 6 498
AK019033.1 - 5, 6 497
BC113760.1 - 5, 6 513 40055351
BM055959.1 - 3, 4 328

View Full Product Details