Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001160163.1 | NP_001153635.1 | 4 | 1287 | |
NM_001160164.1 | NP_001153636.1 | 3 | 1185 | ||
NM_001160165.1 | NP_001153637.1 | 3 | 1313 | ||
NM_015750.3 | NP_056565.1 | 3 | 1153 | ||
GenBank mRNA | AB028023.1 | - | 3 | 1155 | |
AB048604.1 | - | 3 | 1410 | ||
AF139059.1 | - | 3 | 1263 | ||
AK009828.1 | - | 2 | 1345 | ||
AK145284.1 | - | 3 | 1186 | ||
BC026408.1 | - | 2 | 941 | ||
BC105657.1 | - | 3 | 1155 | 40046586 | |
BC105658.1 | - | 3 | 1155 | 40046587 | |
BC105659.1 | - | 3 | 1155 | 40046588 | |
BC105661.1 | - | 2 | 1415 | 40046584 |