Gene Transcripts

Gene Symbol
Pkd2l2
Entrez Gene ID
53871
Gene Name
polycystic kidney disease 2-like 2
Gene Aliases
TRPP5
Chromosome Location
Chr. 18: 34409152 - 34442789 on Build GRCm38
UniGene ID
Mm.445792
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163004.1 NP_001156476.1 14 1864
NM_016927.3 NP_058623.2 14 1864
GenBank mRNA AF182033.1 - 14 1864
AK030120.1 - 1731
AK133423.1 - 14 1861
BC138717.1 - 14 1849 8861739

View Full Product Details