Gene Transcripts

Gene Symbol
Mtmr1
Entrez Gene ID
53332
Gene Name
myotubularin related protein 1
Gene Aliases
AW049210
Chromosome Location
Chr. X: 71364737 - 71419197 on Build GRCm38
UniGene ID
Mm.219672
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016985.2 NP_058681.1 5, 6 706
GenBank mRNA AF073997.1 - 5, 6 679
BC056376.1 - 5, 6 706 6849757
BC057337.1 - 5, 6 700 6825916

View Full Product Details