Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019550.2 | NP_062423.1 | 4 | 439 | |
GenBank mRNA | AF095718.2 | - | 4 | 309 | |
AK041655.1 | - | 4 | 338 | ||
AK050624.1 | - | 4 | 320 | ||
AK050650.1 | - | 4 | 320 | ||
AK135738.1 | - | 4 | 440 | ||
AK137348.1 | - | 4 | 318 | ||
AK207246.1 | - | 4 | 270 | ||
BC010255.1 | - | 4 | 325 | 3709255 |