Gene Transcripts
- Gene Symbol
- Mrps31
- Entrez Gene ID
- 57312
- Gene Name
- mitochondrial ribosomal protein S31
- Gene Aliases
- 1500002D03Rik, AA960481, Imogen38, Imogen44
- Chromosome Location
- Chr. 8: 22410472 - 22431044 on Build GRCm38
- UniGene ID
- Mm.289402
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020560.2 |
NP_065585.1
|
1, 2 |
253 |
|
GenBank mRNA |
AK005110.1 |
- |
1, 2 |
212 |
|
AK011825.1 |
- |
1, 2 |
209 |
|
AK019202.1 |
- |
1, 2 |
233 |
|
AK078073.1 |
- |
1, 2 |
196 |
|
AK161913.1 |
- |
1, 2 |
196 |
|
BC006764.1 |
- |
1, 2 |
148 |
|
BC018475.1 |
- |
1, 2 |
152 |
3492633 |
BC027430.1 |
- |
1, 2 |
157 |
4187438 |
BY237441.1 |
- |
1, 2 |
255 |
|
Z46966.1 |
- |
1, 2 |
172 |
|
View Full Product Details