Gene Transcripts
- Gene Symbol
- Maea
- Entrez Gene ID
- 59003
- Gene Name
- macrophage erythroblast attacher
- Gene Aliases
- 1110030D19Rik, EMP, Gid9
- Chromosome Location
- Chr. 5: 33335572 - 33373294 on Build GRCm38
- UniGene ID
- Mm.138617
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021500.2 |
NP_067475.2
|
2, 3 |
261 |
|
GenBank mRNA |
AF263247.1 |
- |
2, 3 |
223 |
|
AK007307.1 |
- |
2, 3 |
232 |
|
AK034859.1 |
- |
2, 3 |
260 |
|
AK049424.1 |
- |
2, 3 |
281 |
|
AK075637.1 |
- |
1, 2 |
101 |
|
AK088854.1 |
- |
2, 3 |
269 |
|
AK145800.1 |
- |
2, 3 |
324 |
|
AK152523.1 |
- |
2, 3 |
269 |
|
AK168788.1 |
- |
2, 3 |
287 |
|
AK169149.1 |
- |
2, 3 |
269 |
|
BC039054.1 |
- |
2, 3 |
247 |
2648667 |
BC058687.1 |
- |
2, 3 |
259 |
6411707 |
CX212584.1 |
- |
2, 3 |
261 |
|
View Full Product Details