Gene Transcripts

Gene Symbol
Smu1
Entrez Gene ID
74255
Gene Name
smu-1 suppressor of mec-8 and unc-52 homolog (C. elegans)
Gene Aliases
2600001O03Rik, 2610203K23Rik, AB044414, AI845086, AW556129, Bwd
Chromosome Location
Chr. 4: 40735649 - 40757885 on Build GRCm38
UniGene ID
Mm.289929
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021535.4 NP_067510.3 2 274
GenBank mRNA AB044414.1 - 2 203
AK003493.1 - 2 261
AK011140.1 - 2 273
AK019174.1 - 2 233
AK145982.1 - 2 273
AK159635.1 - 2 261
AK168813.1 - 2 268
BC020034.1 - 2 216
BC057446.1 - 2 235 3155760
BY036103.1 - 2 276

View Full Product Details