Gene Transcripts
- Gene Symbol
- Morf4l1
- Entrez Gene ID
- 21761
- Gene Name
- mortality factor 4 like 1
- Gene Aliases
- MORFRG15, MRG15, TEG-189, Tex189, mKIAA4002
- Chromosome Location
- Chr. 9: 90091669 - 90114820 on Build GRCm38
- UniGene ID
- Mm.426209
- Species
- Mus musculus
- Gene Symbol
- Morf4l1b
- Entrez Gene ID
- 627352
- Gene Name
- mortality factor 4 like 1B
- Gene Aliases
- EG627352, Gm6747
- Chromosome Location
- Chr. 19: 41525252 - 41526961 on Build GRCm38
- UniGene ID
- Mm.395111
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK189143.1 |
- |
1 |
280 |
|
View Full Product Details